2-Methyl-1-(2-piperidinylmethyl)piperidine 95%
Fornitore: Apollo Scientific
OR903135-1G
OR903135-250MG
APOSOR903135-1GEA
328
EUR
APOSOR903135-1G
APOSOR903135-250MG
2-Methyl-1-(2-piperidinylmethyl)piperidine 95%
2-Methyl-1-(2-piperidinylmethyl)piperidine
Formula:
C₁₂H₂₄N₂ Peso molecolare: 196,332 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD06653470 CAS Number: 881041-21-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...