Stampa…

[2-(Methylsulphanyl)phenyl]methanol 98%

Fornitore: Apollo Scientific

OR900701-1G OR900701-250MG OR900701-5G
APOSOR900701-1GEA 347 EUR
APOSOR900701-1G APOSOR900701-250MG APOSOR900701-5G
[2-(Methylsulphanyl)phenyl]methanol 98%
[2-(Methylsulphanyl)phenyl]methanol
Formula: C₈H₁₀OS
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00196825
CAS Number: 33384-77-9

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR