3-Hydroxy-N-methylbenzamide 96%
Fornitore: Apollo Scientific
Formula:
C₈H₉NO₂ Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD08729289 CAS Number: 15788-97-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...