1-Chloro-4-(1,3-dibromopropyl)benzene
Fornitore: Apollo Scientific
Formula:
C₉H₉Br₂Cl Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD18311887 CAS Number: 19714-76-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...