Stampa…

4-Amino-4'-methoxybiphenyl hydrochloride

Fornitore: Apollo Scientific
Attenzione

Synonyms: 4'-Methoxy-[1,1'-biphenyl]-4-amine hydrochloride, 4-(4-Methoxyphenyl)aniline hydrochloride

OR7351-1G OR7351-250MG
APOSOR7351-1GEA 98.9 EUR
APOSOR7351-1G APOSOR7351-250MG
4-Amino-4'-methoxybiphenyl hydrochloride
4-Amino-4'-methoxybiphenyl hydrochloride
Formula: C₁₃H₁₃NO·ClH
Peso molecolare: 235,709 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD08445549
CAS Number: 13219-33-5

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR