Chlorin e6
Fornitore: Apollo Scientific
|
Attenzione
|
OR61545-100MG
OR61545-1G
OR61545-250MG
APOSOR61545-100MGEA
187
EUR
APOSOR61545-100MG
APOSOR61545-1G
APOSOR61545-250MG
Chlorin e6
Chlorin e6
Formula:
C₃₄H₃₆N₄O₆ Peso molecolare: 596,673 g/mol Temperatura di stoccaggio: Congelamento |
MDL Number:
MFCD08669566 CAS Number: 19660-77-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...