(1H-Indazol-6-yl)acetic acid
Fornitore: Apollo Scientific
Formula:
C₉H₈N₂O₂ Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD20483631 CAS Number: 221681-76-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...