6-Chloro-4-methylnicotinonitrile
Fornitore: Apollo Scientific
|
Attenzione
|
Synonyms:
2-Chloro-5-cyano-4-methylpyridine, 6-Chloro-4-methylpyridine-3-carbonitrile
OR61017-1G
OR61017-500MG
APOSOR61017-1GEA
274
EUR
APOSOR61017-1G
APOSOR61017-500MG
6-Chloro-4-methylnicotinonitrile
6-Chloro-4-methylnicotinonitrile
Formula:
C₇H₅ClN₂ Peso molecolare: 152,581 g/mol Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD17214204 CAS Number: 66909-35-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...