1-(3-Cyclopropylphenyl)ethanone
Fornitore: Apollo Scientific
|
Attenzione
|
OR55669-1G
OR55669-250MG
APOSOR55669-1GEA
183
EUR
APOSOR55669-1G
APOSOR55669-250MG
1-(3-Cyclopropylphenyl)ethanone
1-(3-Cyclopropylphenyl)ethanone
Formula:
C₁₁H₁₂O Peso molecolare: 160,212 g/mol Temperatura di stoccaggio: Congelamento |
MDL Number:
MFCD06802407 CAS Number: 408359-52-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...