Pentachlorophenyl dodecanoate 97%
Fornitore: Apollo Scientific
|
Pericolo
|
Formula:
C₁₈H₂₃Cl₅O₂ Punto di fusione: 63…65 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00050976 CAS Number: 3772-94-9 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...