N-(2-aminoethyl)prop-2-enamide hydrochloride
Fornitore: Apollo Scientific
Synonyms:
N-(2-Aminoethyl)acrylamide hydrochloride, hydrochloride, N-(2-Aminoethyl)prop-2-enamide
OR55289-250MG
OR55289-1G
APOSOR55289-250MGEA
108
EUR
APOSOR55289-250MG
APOSOR55289-1G
N-(2-aminoethyl)prop-2-enamide hydrochloride
N-(2-aminoethyl)prop-2-enamide hydrochloride
Formula:
C₅H₁₀N₂O·ClH Peso molecolare: 150,607 g/mol Punto di fusione: 120…125 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD18380727 CAS Number: 54641-27-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...