2-Bromo-6-nitrobenzyl bromide
Fornitore: Apollo Scientific
|
Pericolo
|
Synonyms:
1-Bromo-2-(bromomethyl)-3-nitrobenzene
OR55196-1G
OR55196-250MG
OR55196-5G
APOSOR55196-1GEA
56.8
EUR
APOSOR55196-1G
APOSOR55196-250MG
APOSOR55196-5G
2-Bromo-6-nitrobenzyl bromide
2-Bromo-6-nitrobenzyl bromide
Formula:
C₇H₅Br₂NO₂ Peso molecolare: 294,928 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD11036286 CAS Number: 58579-54-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...