5-Bromo-7-chloro-benzofuran-2-carboxylic acid ethyl ester
Fornitore: Apollo Scientific
|
Attenzione
|
Formula:
C₁₁H₈BrClO₃ Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD31544763 CAS Number: 1643914-32-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...