Stampa…

4-Iodo-7-azaindoline, NBOC protected 95%

Fornitore: Apollo Scientific
Attenzione

OR54643-1G OR54643-250MG
APOSOR54643-1GEA 696 EUR
APOSOR54643-1G APOSOR54643-250MG
4-Iodo-7-azaindoline, NBOC protected 95%
4-Iodo-7-azaindoline, NBOC protected
Formula: C₁₂H₁₅IN₂O₂
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD31714176
CAS Number: 2367002-59-1

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR