2,5-Dimethylmorpholine 96%
Fornitore: Apollo Scientific
Formula:
C₆H₁₃NO Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD09971504 CAS Number: 106-56-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...