5-Bromo-1,3-benzenedicarbonyl dichloride 98%
Fornitore: Apollo Scientific
|
Pericolo
|
Formula:
C₈H₃BrCl₂O₂ Punto di fusione: 188…189 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD28009845 CAS Number: 57863-69-1 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...