2-[(Tetrahydro-2H-pyran-4-yl)oxy]benzoyl chloride 97%
Fornitore: Apollo Scientific
|
Pericolo
|
Synonyms:
2-[(Oxan-4-yl)oxy]benzoyl chloride, 4-{[2-(Chlorocarbonyl)phenyl]oxy}tetrahydro-2H-pyran
OR5219-1G
OR5219-250MG
APOSOR5219-1GEA
201
EUR
APOSOR5219-1G
APOSOR5219-250MG
2-[(Tetrahydro-2H-pyran-4-yl)oxy]benzoyl chloride 97%
2-[(Tetrahydro-2H-pyran-4-yl)oxy]benzoyl chloride
Formula:
C₁₂H₁₃ClO₃ Peso molecolare: 240,683 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD09025870 CAS Number: 906352-68-9 UN: 3129 ADR: 4.3+8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...