Stampa…

Methyl 2,4-dihydroxypyrimidine-5-carboxylate

Fornitore: Apollo Scientific
Attenzione

Synonyms: Methyl 2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-carboxylate

OR51846-1G OR51846-5G
APOSOR51846-1GEA 61.1 EUR
APOSOR51846-1G APOSOR51846-5G
Methyl 2,4-dihydroxypyrimidine-5-carboxylate
Methyl 2,4-dihydroxypyrimidine-5-carboxylate
Formula: C₆H₆N₂O₄
Peso molecolare: 170,123 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD25542372
CAS Number: 42821-92-1

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR