Stampa…

Methyl 3-bromo-2-phenyl-propanoate 95%

Fornitore: Apollo Scientific

OR480280-1G OR480280-250MG OR480280-500MG
APOSOR480280-1GEA 641 EUR
APOSOR480280-1G APOSOR480280-250MG APOSOR480280-500MG
Methyl 3-bromo-2-phenyl-propanoate 95%
Methyl 3-bromo-2-phenyl-propanoate
Formula: C₁₀H₁₁BrO₂
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD17215983
CAS Number: 99070-19-6

Visualizza i codici

Specifiche

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR