Stampa…

2-Tetrahydrofuran-2-ylacetic acid

Fornitore: Apollo Scientific

Synonyms: 2-(Tetrahydrofuran-2-yl)acetic acid

OR480227-1G OR480227-250MG OR480227-500MG
APOSOR480227-1GEA 570 EUR
APOSOR480227-1G APOSOR480227-250MG APOSOR480227-500MG
2-Tetrahydrofuran-2-ylacetic acid
2-Tetrahydrofuran-2-ylacetic acid
Formula: C₆H₁₀O₃
Peso molecolare: 130,142 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD03990566
CAS Number: 2434-00-6

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR