t-Butyl 3-(hydroxypropoxyl)-propanoate
Fornitore: Apollo Scientific
Synonyms:
tert-Butyl 3-(3-hydroxypropoxy)propanoate
Formula:
C₁₀H₂₀O₄ Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD30730362 CAS Number: 2100306-78-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...