N-[1-(2-Bromophenyl)ethyl]prop-2-yn-1-amine hydrochloride
Fornitore: Apollo Scientific
|
Attenzione
|
OR475080-1G
OR475080-250MG
APOSOR475080-1GEA
158
EUR
APOSOR475080-1G
APOSOR475080-250MG
N-[1-(2-Bromophenyl)ethyl]prop-2-yn-1-amine hydrochloride
N-[1-(2-Bromophenyl)ethyl]prop-2-yn-1-amine hydrochloride
Formula:
C₁₁H₁₂BrN·ClH Peso molecolare: 274,585 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD31695702 CAS Number: 2366997-03-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...