Stampa…

1-Phenyl-1H-pyrazole-5-boronic acid pinacol ester 97%

Fornitore: Apollo Scientific

Synonyms: 1-Phenyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

OR471708-1G OR471708-250MG
APOSOR471708-1GEA 129 EUR
APOSOR471708-1G APOSOR471708-250MG
1-Phenyl-1H-pyrazole-5-boronic acid pinacol ester 97%
1-Phenyl-1H-pyrazole-5-boronic acid pinacol ester
Formula: C₁₅H₁₉BN₂O₂
Peso molecolare: 270,135 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD23135814
CAS Number: 1238702-58-3

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR