Stampa…

Methyl 1-Boc-3-bromoindole-4-carboxylate

Fornitore: Apollo Scientific

OR471580-1G OR471580-5G
APOSOR471580-1GEA 245 EUR
APOSOR471580-1G APOSOR471580-5G
Methyl 1-Boc-3-bromoindole-4-carboxylate
Methyl 1-Boc-3-bromoindole-4-carboxylate
Formula: C₁₅H₁₆BrNO₄
Peso molecolare: 354,196 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD29065909
CAS Number: 2006277-62-7

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR