1,3-Dimethyl-6-hydroxyquinazoline-2,4-dione
Fornitore: Apollo Scientific
OR471491-1G
OR471491-250MG
OR471491-5G
APOSOR471491-1GEA
448
EUR
APOSOR471491-1G
APOSOR471491-250MG
APOSOR471491-5G
1,3-Dimethyl-6-hydroxyquinazoline-2,4-dione
1,3-Dimethyl-6-hydroxyquinazoline-2,4-dione
Formula:
C₁₀H₁₀N₂O₃ Peso molecolare: 206,198 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD24729909 CAS Number: 1267663-32-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...