6-Chloro-7-methyl-7H-purin-8(9H)-one
Fornitore: Apollo Scientific
OR471475-1G
OR471475-250MG
APOSOR471475-1GEA
916
EUR
APOSOR471475-1G
APOSOR471475-250MG
6-Chloro-7-methyl-7H-purin-8(9H)-one
6-Chloro-7-methyl-7H-purin-8(9H)-one
Formula:
C₆H₅ClN₄O Peso molecolare: 184,583 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD24615587 CAS Number: 1226804-17-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...