5-(Aminomethyl)benzoxazole hydrochloride
Fornitore: Apollo Scientific
|
Attenzione
|
OR471327-1G
OR471327-5G
APOSOR471327-1GEA
611
EUR
APOSOR471327-1G
APOSOR471327-5G
5-(Aminomethyl)benzoxazole hydrochloride
5-(Aminomethyl)benzoxazole hydrochloride
Formula:
C₈H₈N₂O·ClH Peso molecolare: 184,623 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD28895151 CAS Number: 2006277-35-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...