Stampa…

2-Cyano-3-(2-thienyl)propanoic acid

Fornitore: Apollo Scientific

OR471305-1G OR471305-250MG
APOSOR471305-1GEA 601 EUR
APOSOR471305-1G APOSOR471305-250MG
2-Cyano-3-(2-thienyl)propanoic acid
2-Cyano-3-(2-thienyl)propanoic acid
Formula: C₈H₇NO₂S
Peso molecolare: 181,212 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD11593004
CAS Number: 148674-60-6

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR