13-Tetradecynoic acid
Fornitore: Apollo Scientific
OR471273-1G
OR471273-250MG
APOSOR471273-1GEA
1120
EUR
APOSOR471273-1G
APOSOR471273-250MG
13-Tetradecynoic acid
13-Tetradecynoic acid
Formula:
C₁₄H₂₄O₂ Peso molecolare: 224,339 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD18382104 CAS Number: 82909-47-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...