2-Chloro-4-isopropoxypyrimidine
Fornitore: Apollo Scientific
Formula:
C₇H₉ClN₂O Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD16712382 CAS Number: 1250967-81-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...