2-Amino-6-bromobenzamide
Fornitore: Apollo Scientific
Formula:
C₇H₇BrN₂O Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD18378387 CAS Number: 1108790-90-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...