3-(Hydroxymethyl)-2-methylphenylboronic acid pinacol ester
Fornitore: Apollo Scientific
OR471012-1G
OR471012-250MG
OR471012-5G
APOSOR471012-1GEA
916
EUR
APOSOR471012-1G
APOSOR471012-250MG
APOSOR471012-5G
3-(Hydroxymethyl)-2-methylphenylboronic acid pinacol ester
3-(Hydroxymethyl)-2-methylphenylboronic acid pinacol ester
Formula:
C₁₄H₂₁BO₃ Peso molecolare: 248,126 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD18729960 CAS Number: 1400755-05-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...