3-(4-Bromophenyl)cyclobutanol
Fornitore: Apollo Scientific
OR470452-1G
OR470452-250MG
APOSOR470452-1GEA
561
EUR
APOSOR470452-1G
APOSOR470452-250MG
3-(4-Bromophenyl)cyclobutanol
3-(4-Bromophenyl)cyclobutanol
Formula:
C₁₀H₁₁BrO Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD12152571 CAS Number: 1183047-51-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...