Stampa…

Methyl 2-(Boc-amino)-2-(3-piperidyl)acetate

Fornitore: Apollo Scientific

OR470387-1G OR470387-250MG
APOSOR470387-1GEA 1330 EUR
APOSOR470387-1G APOSOR470387-250MG
Methyl 2-(Boc-amino)-2-(3-piperidyl)acetate
Methyl 2-(Boc-amino)-2-(3-piperidyl)acetate
Formula: C₁₃H₂₄N₂O₄
Peso molecolare: 272,341 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD04115494
CAS Number: 1219331-78-8

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR