Stampa…

5-(3-Chlorophenyl)-3-pyridinecarbaldehyde

Fornitore: Apollo Scientific

OR470385-1G OR470385-5G
APOSOR470385-1GEA 499 EUR
APOSOR470385-1G APOSOR470385-5G
5-(3-Chlorophenyl)-3-pyridinecarbaldehyde
5-(3-Chlorophenyl)-3-pyridinecarbaldehyde
Formula: C₁₂H₈ClNO
Peso molecolare: 217,651 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD06410223
CAS Number: 887973-60-6

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR