5-Bromo-2-[(cyclopropylcarbonyl)amino]benzoic acid
Fornitore: Apollo Scientific
Formula:
C₁₁H₁₀BrNO₃ Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD02767110 CAS Number: 401462-74-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...