Stampa…

Methyl 1-(Dimethylcarbamoyl)piperidine-3-carboxylate

Fornitore: Apollo Scientific

OR470209-1G OR470209-5G
APOSOR470209-1GEA 194 EUR
APOSOR470209-1G APOSOR470209-5G
Methyl 1-(Dimethylcarbamoyl)piperidine-3-carboxylate
Methyl 1-(Dimethylcarbamoyl)piperidine-3-carboxylate
Formula: C₁₀H₁₈N₂O₃
Peso molecolare: 214,262 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD18651717
CAS Number: 1272756-08-7

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR