2-Chloro-3-nitrobenzamide
Fornitore: Apollo Scientific
OR470205-1G
OR470205-5G
APOSOR470205-1GEA
183
EUR
APOSOR470205-1G
APOSOR470205-5G
2-Chloro-3-nitrobenzamide
2-Chloro-3-nitrobenzamide
Formula:
C₇H₅ClN₂O₃ Peso molecolare: 200,579 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00482025 CAS Number: 117054-76-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...