Stampa…

4-[2-(Chloromethyl)benzimidazol-1-yl]butyl Pivalate

Fornitore: Apollo Scientific

OR470178-1G OR470178-5G
APOSOR470178-1GEA 459 EUR
APOSOR470178-1G APOSOR470178-5G
4-[2-(Chloromethyl)benzimidazol-1-yl]butyl Pivalate
4-[2-(Chloromethyl)benzimidazol-1-yl]butyl Pivalate
Formula: C₁₇H₂₃ClN₂O₂
Peso molecolare: 322,83 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD18651695
CAS Number: 807316-60-5

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR