2-[4-(Methylthio)phenoxy]acetonitrile
Fornitore: Apollo Scientific
OR470162-25G
OR470162-5G
APOSOR470162-25GEA
713
EUR
APOSOR470162-25G
APOSOR470162-5G
2-[4-(Methylthio)phenoxy]acetonitrile
2-[4-(Methylthio)phenoxy]acetonitrile
Formula:
C₉H₉NOS Peso molecolare: 179.239 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD18071041 CAS Number: 43111-34-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...