3-Bromo-4-pyrrolidinobenzoic acid
Fornitore: Apollo Scientific
OR470062-1G
OR470062-5G
APOSOR470062-1GEA
256
EUR
APOSOR470062-1G
APOSOR470062-5G
3-Bromo-4-pyrrolidinobenzoic acid
3-Bromo-4-pyrrolidinobenzoic acid
Formula:
C₁₁H₁₂BrNO₂ Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD11110874 CAS Number: 1131615-12-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...