Stampa…

2-Piperidylacetic acid hydrochloride

Fornitore: Apollo Scientific
Pericolo

Synonyms: 2-Piperidylacetic Acid Hydrochloride

OR470052-1G OR470052-5G
APOSOR470052-1GEA 154 EUR
APOSOR470052-1G APOSOR470052-5G
2-Piperidylacetic acid hydrochloride
2-Piperidylacetic acid hydrochloride
Formula: C₇H₁₃NO₂·ClH
Peso molecolare: 179,645 g/mol
Punto di fusione: 183…186 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD08061346
CAS Number: 19615-30-6
UN: 1759
ADR: 8,III

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR