2-Amino-5-bromo-1H-indole hydrochloride
Fornitore: Apollo Scientific
|
Attenzione
|
OR46766-1G
OR46766-250MG
APOSOR46766-1GEA
654
EUR
APOSOR46766-1G
APOSOR46766-250MG
2-Amino-5-bromo-1H-indole hydrochloride
2-Amino-5-bromo-1H-indole hydrochloride
Formula:
C₈H₇BrN₂·ClH Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD24570261 CAS Number: 28493-02-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...