7-Benzyloxy-4-chloro-6-methoxyquinoline
Fornitore: Apollo Scientific
Synonyms:
7-(Benzyloxy)-4-chloro-6-methoxyquinoline
Formula:
C₁₇H₁₄ClNO₂ Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD12910463 CAS Number: 286371-49-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...