2,2,3,3,5,5,6-Heptachloro-1,4-dioxane
Fornitore: Apollo Scientific
|
Attenzione
|
OR460053-1G
OR460053-5G
APOSOR460053-1GEA
147
EUR
APOSOR460053-1G
APOSOR460053-5G
2,2,3,3,5,5,6-Heptachloro-1,4-dioxane
2,2,3,3,5,5,6-Heptachloro-1,4-dioxane
Formula:
C₄HCl₇O₂ Peso molecolare: 329,221 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00085459 CAS Number: 6629-96-5 UN: 1165 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...