(E)-1,2-Dichloropropene 97%
Fornitore: Apollo Scientific
|
Attenzione
|
Formula:
C₃H₄Cl₂ Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00040138 CAS Number: 7069-38-7 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...