Stampa…

b-Amino-3-bromobenzenepropanol 97%

Fornitore: Apollo Scientific

OR452176-1G OR452176-250MG OR452176-500MG
APOSOR452176-1GEA 783 EUR
APOSOR452176-1G APOSOR452176-250MG APOSOR452176-500MG
b-Amino-3-bromobenzenepropanol 97%
b-Amino-3-bromobenzenepropanol
Formula: C₉H₁₂BrNO
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD10034636
CAS Number: 254427-26-4

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR