Stampa…

2-Bromo-5-(cyclopropyl)thiophene-3-carboxylic acid

Fornitore: Apollo Scientific

OR45207-1G OR45207-250MG OR45207-5G
APOSOR45207-1GEA 575 EUR
APOSOR45207-1G APOSOR45207-250MG APOSOR45207-5G
2-Bromo-5-(cyclopropyl)thiophene-3-carboxylic acid
2-Bromo-5-(cyclopropyl)thiophene-3-carboxylic acid
Formula: C₈H₇BrO₂S
Peso molecolare: 247,109 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD30003636
CAS Number: 2091719-00-3

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR