4-Bromo-L-tryptophan 97%
Fornitore: Apollo Scientific
OR452027-1G
OR452027-250MG
OR452027-500MG
APOSOR452027-1GEA
980
EUR
APOSOR452027-1G
APOSOR452027-250MG
APOSOR452027-500MG
4-Bromo-L-tryptophan 97%
4-Bromo-L-tryptophan
Formula:
C₁₁H₁₁BrN₂O₂ Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD10565749 CAS Number: 52448-16-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...