Stampa…

7-Bromo-L-tryptophan 97%

Fornitore: Apollo Scientific

Synonyms: H-Trp(7-Br)-OH, (S)-2-Amino-3-(7-bromo-1H-indol-3-yl)propanoic acid

OR452008-250MG OR452008-500MG
APOSOR452008-250MGEA 700 EUR
APOSOR452008-250MG APOSOR452008-500MG
7-Bromo-L-tryptophan 97%
7-Bromo-L-tryptophan
Formula: C₁₁H₁₁BrN₂O₂
Peso molecolare: 283,121 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD09264335
CAS Number: 75816-19-2

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR